Merck
  • Sustainable Plastics from Biomass: Blends of Polyesters Based on 2,5-Furandicarboxylic Acid.
Sustainable Plastics from Biomass: Blends of Polyesters Based on 2,5-Furandicarboxylic Acid.
Polymers (2020-01-23)
Niki Poulopoulou, Dimitra Smyrnioti, George N Nikolaidis, Ilektra Tsitsimaka, Evi Christodoulou, Dimitrios N Bikiaris, Maria Anna Charitopoulou, Dimitris S Achilias, Maria Kapnisti, George Z Papageorgiou
ABSTRACT
Intending to expand the thermo-physical properties of bio-based polymers, furan-based thermoplastic polyesters were synthesized following the melt polycondensation method. The resulting polymers, namely, poly(ethylene 2,5-furandicarboxylate) (PEF), poly(propylene 2,5-furandicarboxylate) (PPF), poly(butylene 2,5-furandicarboxylate) (PBF) and poly(1,4-cyclohexanedimethylene 2,5-furandicarboxylate) (PCHDMF) are used in blends together with various polymers of industrial importance, including poly(ethylene terephthalate) (PET), poly(ethylene 2,6-naphthalate) (PEN), poly(L-lactic acid) (PLA) and polycarbonate (PC). The blends are studied concerning their miscibility, crystallization and solid-state characteristics by using wide-angle X-ray diffractometry (WAXD), differential scanning calorimetry (DSC) and polarized light microscopy (PLM). PEF blends show in general dual glass transitions in the DSC heating traces for the melt quenched samples. Only PPF-PEF blends show a single glass transition and a single melt phase in PLM. PPF forms immiscible blends except with PEF and PBF. PBF forms miscible blends with PCHDMF and PPF, whereas all other blends show dual glass transitions in DSC and phase separation in PLM. PCHDMF-PEF and PEN-PEF blends show two glass transition temperatures, but they shift to intermediate temperature values depending on the composition, indicating some partial miscibility of the polymer pairs.