Merck
  • Stretchable Lithium-Ion Battery Based on Re-entrant Micro-honeycomb Electrodes and Cross-Linked Gel Electrolyte.
Stretchable Lithium-Ion Battery Based on Re-entrant Micro-honeycomb Electrodes and Cross-Linked Gel Electrolyte.
ACS nano (2020-03-03)
Seulki Kang, Soo Yeong Hong, Nayeon Kim, Jinwoo Oh, Min Park, Kyung Yoon Chung, Sang-Soo Lee, Jonghwi Lee, Jeong Gon Son
ABSTRACT
Stretchable energy storage devices are of great interest because of their potential applications in body-friendly, skin-like, wearable devices. However, stretchable batteries are very challenging to fabricate. The electrodes must have a degree of stretchability because the active materials occupy most of the volume, and the separator and packaging should also be stretchable. Here, an all-component stretchable lithium-ion battery was realized by leveraging the structural stretchability of re-entrant micro-honeycomb graphene-carbon nanotube (CNT)/active material composite electrodes and a physically cross-linked gel electrolyte, without using an inactive elastomeric substrate or matrix. Active materials interconnected via the entangled CNT and graphene sheets provided a mechanically stable porous network framework, and the inwardly protruding framework in the re-entrant honeycomb structure allowed for structural stretching during deformation. The composite network consisting solely of binder-free, highly conductive materials provided superior electron transfer, and vertically aligned microchannels enabled facile ion transport. Additionally, the physically cross-linked gel electrolyte increased the mechanical stability upon deformation of the electrodes and was effective as a stretchable separator. The resulting stretchable battery showed a high areal capacity of 5.05 mAh·cm-2, superior electrochemical performance up to 50% strain under repeated (up to 500) stretch-release cycles, and long-term stability of 95.7% after 100 cycles in air conditions.