Merck
Shock Exfoliation of Graphene Fluoride in Microwave.
Small (Weinheim an der Bergstrasse, Germany) (2019-09-10)
Sicheng Wu, Jingxin Mo, Yachao Zeng, Yuan Wang, Aditya Rawal, Jason Scott, Zhen Su, Wenhao Ren, Sheng Chen, Kaixuan Wang, Wei Chen, Yongzhi Zhang, Chuan Zhao, Xianjue Chen
ABSTRACT
An unprecedented microwave-based strategy is developed to facilitate solid-phase, instantaneous delamination and decomposition of graphite fluoride (GF) into few-layer, partially fluorinated graphene. The shock reaction occurs (and completes in few seconds) under microwave irradiation upon exposing GF to either "microwave-induced plasma" generated in vacuum or "catalyst effect" caused by intense sparking of graphite at ambient conditions. A detailed analysis of the structural and compositional transformations in these processes indicates that the GF experiences considerable exfoliation and defluorination, during which sp2 -bonded carbon is partially recovered despite significant structural defects being introduced. The exfoliated fluorinated graphene shows excellent electrochemical performance as anode materials in potassium ion batteries and as catalysts for the conversion of O2 to H2 O2 . This simple and scalable method requires minimal energy input and does not involve the use of other chemicals, which is attractive for extensive research in fluorine-containing graphene and its derivatives in laboratories and industrial applications.