Merck
  • Beta-1,3-glucanase from Delftia tsuruhatensis strain MV01 and its potential application in vinification.
Beta-1,3-glucanase from Delftia tsuruhatensis strain MV01 and its potential application in vinification.
Applied and environmental microbiology (2010-12-21)
V Blättel, M Larisika, P Pfeiffer, C Nowak, A Eich, J Eckelt, H König
ABSTRACT
During vinification microbial activities can spoil wine quality. As the wine-related lactic acid bacterium Pediococcus parvulus is able to produce slimes consisting of a β-1,3-glucan, must and wine filtration can be difficult or impossible. In addition, the metabolic activities of several wild-type yeasts can also negatively affect wine quality. Therefore, there is a need for measures to degrade the exopolysaccharide from Pediococcus parvulus and to inhibit the growth of certain yeasts. We examined an extracellular β-1,3-glucanase from Delftia tsuruhatensis strain MV01 with regard to its ability to hydrolyze both polymers, the β-1,3-glucan from Pediococcus and that from yeast cell walls. The 29-kDa glycolytic enzyme was purified to homogeneity. It exhibited an optimal activity at 50°C and pH 4.0. The sequencing of the N terminus revealed significant similarities to β-1,3-glucanases from different bacteria. In addition, the investigations indicated that this hydrolytic enzyme is still active under wine-relevant parameters such as elevated ethanol, sulfite, and phenol concentrations as well as at low pH values. Therefore, the characterized enzyme seems to be a useful tool to prevent slime production and undesirable yeast growth during vinification.