Merck
  • Electrochemical detection of quorum sensing signaling molecules by dual signal confirmation at microelectrode arrays.
Electrochemical detection of quorum sensing signaling molecules by dual signal confirmation at microelectrode arrays.
Analytical chemistry (2011-02-18)
Eva Baldrich, Francesc Xavier Muñoz, Cristina García-Aljaro
ABSTRACT
n-Acyl homoserine lactones (AHLs) are produced by gram-negative bacteria to regulate gene expression in a cell density dependent manner. For instance, expression of virulence factors by pathogens such as Pseudomonas aeruginosa is induced only when a threshold concentration of AHLs is reached, which indicates that the bacterial population is big enough to promote infection. In this study, the indicator strain Agrobacterium tumefaciens NTL4 (pZLR4), which carries a β-galactosidase (β-gal) reporter gene under the control of a quorum sensing promoter, was used to develop an electrochemical biosensor to detect AHLs using the model n-(3-oxo)-dodecanoyl-L-homoserine lactone (oxo-C12-HSL), an AHL previously detected in cystic fibrosis patients infected with P. aeruginosa. The substrate 4-aminophenyl β-D-galactopyranoside was used to detect β-gal activity by cyclic voltammetry. Furthermore, simultaneous monitoring of substrate consumption and p-aminophenol production by β-gal allowed on-chip result verification by dual-signal confirmation. The sensor exhibited high reproducibility and accurately detected oxo-C12-HSL in a low picomolar to low nanomolar range in spiked liquid cultures and artificial saliva, as well as AHLs naturally released by P. aeruginosa in culture supernatants. Moreover, detection took just 2 h, required no sample pretreatment or preconcentration steps, and was easier and faster than traditional methods.