Merck
Organometallic half-sandwich iridium anticancer complexes.
Journal of medicinal chemistry (2011-03-30)
Zhe Liu, Abraha Habtemariam, Ana M Pizarro, Sally A Fletcher, Anna Kisova, Oldrich Vrana, Luca Salassa, Pieter C A Bruijnincx, Guy J Clarkson, Viktor Brabec, Peter J Sadler
ABSTRACT
The low-spin 5d(6) Ir(III) organometallic half-sandwich complexes [(η(5)-Cp(x))Ir(XY)Cl](0/+), Cp(x) = Cp*, tetramethyl(phenyl)cyclopentadienyl (Cp(xph)), or tetramethyl(biphenyl)cyclopentadienyl (Cp(xbiph)), XY = 1,10-phenanthroline (4-6), 2,2'-bipyridine (7-9), ethylenediamine (10 and 11), or picolinate (12-14), hydrolyze rapidly. Complexes with N,N-chelating ligands readily form adducts with 9-ethylguanine but not 9-ethyladenine; picolinate complexes bind to both purines. Cytotoxic potency toward A2780 human ovarian cancer cells increases with phenyl substitution on Cp*: Cp(xbiph) > Cp(xph) > Cp*; Cp(xbiph) complexes 6 and 9 have submicromolar activity. Guanine residues are preferential binding sites for 4-6 on plasmid DNA. Hydrophobicity (log P), cell and nucleus accumulation of Ir correlate with cytotoxicity, 6 > 5 > 4; they distribute similarly within cells. The ability to displace DNA intercalator ethidium bromide from DNA correlates with cytotoxicity and viscosity of Ir-DNA adducts. The hydrophobicity and intercalative ability of Cp(xph) and Cp(xbiph) make a major contribution to the anticancer potency of their Ir(III) complexes.