Merck
  • Renal expression and urinary excretion of liver-type fatty acid-binding protein in cats with renal disease.
Renal expression and urinary excretion of liver-type fatty acid-binding protein in cats with renal disease.
Journal of veterinary internal medicine (2020-02-23)
Masaaki Katayama, Keiichi Ohata, Tamako Miyazaki, Rieko Katayama, Nobuko Wakamatsu, Misa Ohno, Tetsuro Yamashita, Tsuyoshi Oikawa, Takeshi Sugaya, Masao Miyazaki
ABSTRACT
Liver-type fatty acid-binding protein (L-FABP) is a biomarker for early detection of renal disease in humans. Liver-type fatty acid-binding protein is cytotoxic oxidation products secreted from proximal tubules under ischemia and oxidative stress. To examine renal expression and quantify urinary excretion of L-FABP in catswith renal disease. One hundred and thirty-four client-owned cats including 34 cats with serum creatinine (sCre) values >1.6 mg/dL and 10 other cats that died in clinics. Tissue expressions of L-FABP were examined by reverse transcription polymerase chain reaction and Western blotting. Urinary L-FABP (uL-FABP) and serum L-FABP (sL-FABP) levels were determined by enzyme-linked immunosorbent assay. Anti-liver-type fatty acid-binding protein antibody immunostained renal sections. Feline kidneys express L-FABP. Strong L-FABP signals were observed in the lumens of proximal tubular cells in 5 cats with high uL-FABP excretion, but not in 5 cats with low uL-FABP excretion. In 9 normal cats, uL-FABP index was <1.2 μg/g urinary creatinine (uCre). High uL-FABP indexes (>10.0 μg/g uCre) were detected in 7 of 100 cats with low sCre (<1.6 mg/dL) and 18 of 44 cats with high sCre (>1.6 mg/dL). There was a weak correlation between L-FABP index and sCre, serum symmetric dimethylarginine (SDMA), or blood urea nitrogen (BUN), and these correlation coefficients were increased by analyzing only data of cats with sCre >1.6 mg/dL. There was a weak correlation between u L-FABP index and sL-FABP in all tested cats, but not in cats with high sCre. This study demonstrates correlations between L-FABP and current renal biomarkers for chronic kidney disease in cats, such as sCre and SDMA. Liver-type fatty acid-binding protein may be a potential biomarker to predict early pathophysiological events in feline kidneys.