Merck
  • Residues and enantioselective behavior of cyflumetofen from apple production.
Residues and enantioselective behavior of cyflumetofen from apple production.
Food chemistry (2020-04-04)
Rui Quan, Minmin Li, Yongguo Liu, Nuo Jin, Jia Zhang, Ruixing Li, Fengzhong Wang, Zhidong Wang, Frédéric Francis, Zhiqiang Kong, Bei Fan
ABSTRACT
The effect of food processing on the level and fate of chiral pesticide residues in apple products has rarely been investigated. In this study, we used ultra-performance convergence chromatography coupled with triple quadrupole mass spectrometry to determine the content of the novel chiral acaricide cyflumetofen. The matrix-matched calibration lines were constructed for apple slices, juice, wine and vinegar, and the determination coefficients (r2) exceeded 0.9954. Acceptable average recoveries were within 81.1% to 119.9%, and the relative standard deviations (RSDs) ranged from 0.8% to 11.0%. Processing effectiveness is represented by the processing factor (PF). The results indicated that the PFs of different procedures (washing, peeling, enzymolysis, fermentation, among others.) were generally less than 1. The reduction of cyflumetofen enantiomers during fermentation was in accordance with first-order kinetics, and stereoselective behavior was observed. This study provides reliable references for the risk assessment of cyflumetofen in the processing of apple products.