Merck
Hypoxia induces adenosine release from the rat carotid body.
Journal of neurochemistry (2004-05-19)
Sílvia V Conde, Emília C Monteiro
ABSTRACT
The effect of hypoxia on the release of adenosine was studied in vitro in the rat whole carotid body (CB) and compared with the effect of hypoxia (2%, 5% and 10% O(2)) on adenosine concentrations in superior cervical ganglia (SCG) and carotid arteries. Moderate hypoxia (10% O(2)) increased adenosine concentrations released from the CBs by 44%, but was not a strong enough stimulus to evoke adenosine release from SCG and arterial tissue. The extracellular pathways of adenosine production in rat CBs in normoxia and hypoxia were also investigated. S-(p-nitrobenzyl)-6-thioinosine (NBTI) and dipyridamole were used as pharmacological tools to inhibit adenosine equilibrative transporters (ENT) and alpha,beta-methylene ADP (AOPCP) to inhibit ecto-5'-nucleotidase. Approximately 40% of extracellular adenosine in the CB came from the extracellular catabolism of ATP, under both normoxic and hypoxic conditions. Low pO(2) triggers adenosine efflux through activation of NBTI-sensitive ENT. This effect was only apparent in hypoxia and when adenosine extracellular concentrations were reduced by the blockade of ecto-5'-nucleotidase. We concluded that CB chemoreceptor sensitivity could be related to its low threshold for the release of adenosine in response to hypoxia here quantified for the first time.