Merck
  • Cloning of Protocatechuate 3,4-Dioxygenase Genes from Bradyrhizobium japonicum USDA110.
Cloning of Protocatechuate 3,4-Dioxygenase Genes from Bradyrhizobium japonicum USDA110.
Applied and environmental microbiology (1993-08-01)
G K Podila, S Kotagiri, S Shantharam
ABSTRACT
A heterologous gene probe encoding the alpha and beta subunits of the Pseudomonas cepacia protocatechuate 3,4-dioxygenase (PCD) was used to detect its homolog in the genome of Bradyrhizobium japonicum USDA110. Three cosmid clones carrying a 2.2-kb BamHI insert showed high levels of PCD activity. SacI digestion of one of the genomic clones, pBjG17, produced a 2.5-kb insert DNA that complemented a PCD mutant of P. cepacia.