- Conversion of glycogen phosphorylase b to a by non-activated phosphorylase b kinase: an in vitro model of the mechanism of increase in phosphorylase a activity with muscle contraction.
Conversion of glycogen phosphorylase b to a by non-activated phosphorylase b kinase: an in vitro model of the mechanism of increase in phosphorylase a activity with muscle contraction.
Proceedings of the National Academy of Sciences of the United States of America (1970-09-01)
C Villar-Palasi, S H Wei
PMID4318782
ABSTRACT
Phosphorylase b kinase activity, as present in resting muscle in the non-activated form, appears to be ample to account for the fast appearance of phosphorylase a observed with muscle contraction. The kinase activity is repressed by free ATP and stimulated by free Mg(2+). Phosphorylase b kinase activity increases greatly when the Mg(2+):ATP ration exceeds 1. It is proposed that the breakdown of ATP that occurs during muscle contraction may represent the triggering factor for the observed in vivo conversion of phosphorylase b into a.