Merck
  • Synthetic peptides containing ITIM-like sequences of IREM-1 (CD300F) differentially regulate MyD88 and TRIF-mediated TLR signalling through activation of SHP and/or PI3K.
Synthetic peptides containing ITIM-like sequences of IREM-1 (CD300F) differentially regulate MyD88 and TRIF-mediated TLR signalling through activation of SHP and/or PI3K.
Clinical and experimental immunology (2012-02-01)
S-M Lee, K Suk, W-H Lee
ABSTRACT
The immune receptor expressed on myeloid cells 1 (IREM-1/CD300F) has been shown to inhibit various inflammatory processes in myeloid cells, such as macrophages and mast cells. IREM-1 exerts its inhibitory effect through its intracellular immunoreceptor tyrosine-based inhibition motifs (ITIMs). In order to generate immunomodulatory molecules that can regulate the inflammatory activation of macrophages, decapeptides representing each of the five ITIM-like sequences in the cytoplasmic tail of IREM-1 were synthesized in conjugation with human immunodeficiency virus-transactivator of transcription (HIV-TAT(48-57)), which was added to promote internalization of the peptides. Interestingly, all these TAT-ITIM fusion peptides inhibited Toll-like receptor (TLR)-mediated production of proinflammatory molecules, including matrix metalloproteinase (MMP)-9, tumour necrosis factor (TNF)-α, monocyte chemotactic protein-1 (MCP-1) and interleukin (IL)-8. When various TLR ligands were used to stimulate the human macrophage-like cell line human acute monocytic leukaemia cell line (THP)-1, the TAT-ITIM peptides blocked both myeloid differentiation factor 88 (MyD88) and Toll-interleukin 1 receptor (TIR)-domain-containing adapter-inducing interferon-β (TRIF)-mediated TLR signalling pathways. Utilization of specific inhibitors and detection of the active form of signalling adaptors by Western blot analysis further demonstrated that the inhibitory effects of these TAT-ITIM peptides require activation of Src homology 2 (SH2)-containing tyrosine phosphatase (SHP) and/or phosphoinositide 3-kinase (PI3K). These data indicate that these synthetic peptides may be used to regulate immune responses that involve TLR-mediated inflammatory activation of macrophages.