Merck
  • Structure and mechanism of bactericidal mammalian perforin-2, an ancient agent of innate immunity.
Structure and mechanism of bactericidal mammalian perforin-2, an ancient agent of innate immunity.
Science advances (2020-02-18)
Tao Ni, Fang Jiao, Xiulian Yu, Saša Aden, Lucy Ginger, Sophie I Williams, Fangfang Bai, Vojtěch Pražák, Dimple Karia, Phillip Stansfeld, Peijun Zhang, George Munson, Gregor Anderluh, Simon Scheuring, Robert J C Gilbert
ABSTRACT
Perforin-2 (MPEG1) is thought to enable the killing of invading microbes engulfed by macrophages and other phagocytes, forming pores in their membranes. Loss of perforin-2 renders individual phagocytes and whole organisms significantly more susceptible to bacterial pathogens. Here, we reveal the mechanism of perforin-2 activation and activity using atomic structures of pre-pore and pore assemblies, high-speed atomic force microscopy, and functional assays. Perforin-2 forms a pre-pore assembly in which its pore-forming domain points in the opposite direction to its membrane-targeting domain. Acidification then triggers pore formation, via a 180° conformational change. This novel and unexpected mechanism prevents premature bactericidal attack and may have played a key role in the evolution of all perforin family proteins.