Merck
  • Investigation on Antibiotic-Resistance, Biofilm Formation and Virulence Factors in Multi Drug Resistant and Non Multi Drug Resistant Staphylococcus pseudintermedius.
Investigation on Antibiotic-Resistance, Biofilm Formation and Virulence Factors in Multi Drug Resistant and Non Multi Drug Resistant Staphylococcus pseudintermedius.
Microorganisms (2020-01-01)
Gabriele Meroni, Joel F Soares Filipe, Lorenzo Drago, Piera A Martino
ABSTRACT
Staphylococcus pseudintermedius is a commensal bacterium frequently isolated from canine skin and recognized as a zoonotic agent especially for dog-owners. This study focused on (a) the antibiotic-resistance phenotypes; (b) the ability to produce biofilm (slime); and (c) the dissemination of virulence factors in S. pseudintermedius strains. Seventy-three S. pseudintermedius strains were screened for antibiotic-resistance against 22 different molecules by means of Kirby-Bauer assay. The ability to produce biofilm was investigated using the microtiter plate assay (MtP) and the amplification of icaA and icaD genes. Virulence factors such as cytotoxins (lukI), enterotoxins (seC), and exfoliative toxins (siet, expA, and expB) were evaluated. The antibiotic-resistance profiles revealed 42/73 (57%) multi-drug resistant (MDR) strains and 31/73 (43%) not-MDR. All the MDR strains and 8/31 (27%) of not-MDR resulted in biofilm producers. Leukotoxin LukI was found in 70/73 (96%) of the isolates. Moreover, the enterotoxin gene seC was detected in 47/73 (64%) of the strains. All the isolates carried the siet gene, whereas expA and expB were found in 3/73 (4%) and 5/73 (7%), respectively. In conclusion, S. pseudintermedius should be considered a potential zoonotic and human agent able to carry different virulence determinants and capable of producing biofilm which facilitates horizontal gene transfer.