Merck
  • Defective survival of naive CD8+ T lymphocytes in the absence of the beta3 regulatory subunit of voltage-gated calcium channels.
Defective survival of naive CD8+ T lymphocytes in the absence of the beta3 regulatory subunit of voltage-gated calcium channels.
Nature immunology (2009-10-20)
Mithilesh K Jha, Abdallah Badou, Marcel Meissner, John E McRory, Marc Freichel, Veit Flockerzi, Richard A Flavell
ABSTRACT
The survival of T lymphocytes requires sustained, Ca(2+) influx-dependent gene expression. The molecular mechanism that governs sustained Ca(2+) influx in naive T lymphocytes is unknown. Here we report an essential role for the beta3 regulatory subunit of voltage-gated calcium (Ca(v)) channels in the maintenance of naive CD8(+) T cells. Deficiency in beta3 resulted in a profound survival defect of CD8(+) T cells. This defect correlated with depletion of the pore-forming subunit Ca(v)1.4 and attenuation of T cell antigen receptor (TCR)-mediated global Ca(2+) entry in CD8(+) T cells. Ca(v)1.4 and beta3 associated with T cell signaling machinery and Ca(v)1.4 localized in lipid rafts. Our data demonstrate a mechanism by which Ca(2+) entry is controlled by a Ca(v)1.4-beta3 channel complex in T cells.