Merck
  • Endothelin ETA receptors and endothelium partially mediate the positive inotropic and lusitropic effects of angiotensin II.
Endothelin ETA receptors and endothelium partially mediate the positive inotropic and lusitropic effects of angiotensin II.
European journal of pharmacology (2006-07-18)
Paulo Castro-Chaves, Roberto Roncon-Albuquerque, Adelino F Leite-Moreira
ABSTRACT
We analyzed the influence of endothelin-1 and endocardial endothelium on the myocardial effects of angiotensin-II. Angiotensin-II (10(-9)-10(-5) M) was tested in rabbit right papillary muscles in absence (Protocol-A) or presence of PD-145065 (10(-7) M; Protocol-B), BQ-123 (10(-7) M; Protocol-C) or losartan (10(-6) M; Protocol-E), as well as, after removing the endocardial endothelium with Triton X-100 0.5% (Protocol-D). In Protocol-F increasing concentrations of endothelin-1 (10(-10)-10(-8) M) were added in presence of angiotensin-II (10(-7) M) after selective removal of the endocardial endothelium. In Protocol-A, angiotensin-II had dose-dependent positive inotropic and lusitropic effects, maximal at 10(-6) M increasing 122+/-13% active tension, 117+/-16% dT/dtmax and 86+/-9% dT/dtmin. In Protocols B, C and D the inotropic and lusitropic effects of angiotensin-II were significantly attenuated. The same concentration (10(-6) M) of angiotensin-II increased respectively 48+/-11%, 59+/-27% and 72+/-16% active tension; 54+/-14%, 54+/-20% and 32+/-9% dT/dtmax; and 39+/-8%, 48+/-19% and 59+/-11% dT/dtmin; and 40+/-10%. EC(50) for active tension significantly increased from -7.8+/-0.1 logM in Protocol A to -7.1+/-0.3, -6.7+/-0.4 and -6.8+/-0.3 logM in Protocols B, C and D respectively, while Emax decreased from 106+/-14% in Protocol A to 50+/-14 and 51+/-19% in Protocols B and C respectively, but did not significantly change in Protocol D (114+/-25%). Losartan completely blocked the inotropic and lusitropic effects of angiotensin-II, while the attenuation of these effects after the selective removal of the endocardial endothelium was blunted by concomitant administration of endothelin-1 (Protocol F). In conclusion, angiotensin-II has a dose-dependent positive inotropic effect that depends, to a great extent, on endothelin ETA receptor activation and intact endocardial endothelium.