Merck
  • Membrane receptor-mediated apoptosis and caspase activation in the differentiated EoL-1 eosinophilic cell line.
Membrane receptor-mediated apoptosis and caspase activation in the differentiated EoL-1 eosinophilic cell line.
Journal of leukocyte biology (2004-04-13)
Mohammed W Al-Rabia, Morgan G Blaylock, Darren W Sexton, Garry M Walsh
ABSTRACT
Caspases are key molecules in the control of apoptosis, but relatively little is known about their contribution to eosinophil apoptosis. We examined caspase-3, -8, and -9 activities in receptor ligation-dependent apoptosis induction in the differentiated human eosinophilic cell line EoL-1. Differentiated EoL-1 exhibited bi-lobed nuclei, eosinophil-associated membrane receptors, and basic granule proteins. Annexin-V fluorescein isothiocyanate binding to EoL-1 revealed significant (P<0.01) apoptosis induction in cells cultured for 20 h with monoclonal antibodies (mAb) specific for CD45 (71%+/-4.3), CD45RA (58%+/-2.3), CD45RB (68%+/-2.4), CD95 (47%+/-2.6), and CD69 (52%+/-2.1) compared with control (23%+/-1.6) or CD45RO mAb (27%+/-3.9). The pan-caspase inhibitor Z-Val-Ala-Asp-fluoromethylketone (fmk) and inhibitors of caspase-8 (Z-Ile-Glu-Thr-Asp-fmk) and caspase-9 (Z-Leu-Glu-His-Asp-fmk) significantly inhibited mAb-induced apoptosis of EoL-1 but had no effect on constitutive (baseline) apoptosis at 16 and 20 h. Caspase activity was analyzed using the novel CaspaTag trade mark technique and flow cytometry. EoL-1 treated with pan-CD45, CD45RA, CD45RB, and CD95 mAb exhibited caspase-3 and -9 activation at 12 h post-treatment, which increased at 16 and 20 h. Activated caspase-8 was detected 12 and 16 h after ligation with CD45, CD45RA, CD45RB, and CD95 mAb followed by a trend toward basal levels at 20 h. CD69 ligation resulted in caspase-3 activation, a modest but significant activation of caspase-8, and a loss in mitochondrial transmembrane potential but had no significant effect on activation of caspase-9. Thus, the intrinsic and extrinsic caspase pathways are involved in controlling receptor ligation-mediated apoptosis induction in human eosinophils, findings that may aid the development of a more targeted, anti-inflammatory therapy for asthma.