Merck
  • The furin inhibitor hexa-D-arginine blocks the activation of Pseudomonas aeruginosa exotoxin A in vivo.
The furin inhibitor hexa-D-arginine blocks the activation of Pseudomonas aeruginosa exotoxin A in vivo.
Infection and immunity (2002-11-20)
Miroslav S Sarac, Angus Cameron, Iris Lindberg
ABSTRACT
The Pseudomonas aeruginosa exotoxin A (PEA) protein requires furin-mediated cleavage for manifestation of toxicity. We show here that the small stable furin inhibitor hexa-D-arginine amide effectively blocks PEA-induced cell lysis and is itself noncytotoxic. Administration of hexa-D-arginine to PEA-treated mice significantly improves their survival rate and also decreases circulating levels of tumor necrosis factor alpha.