Merck
  • Comparison of Cu salts and commercial Cu based fungicides on toxicity towards microorganisms in soil.
Comparison of Cu salts and commercial Cu based fungicides on toxicity towards microorganisms in soil.
Environmental pollution (Barking, Essex : 1987) (2019-11-23)
Raquel Vázquez-Blanco, Manuel Arias-Estévez, Erland Bååth, David Fernández-Calviño
ABSTRACT
Microbial responses to Cu pollution as a function of Cu sources (Cu salts and commercial Cu fungicides) were assessed in a soil using basal soil respiration, and bacterial and fungal community growth, as endpoints. The soil was amended with different concentrations (0-32 mmol Cu kg-1) of Cu nitrate, Cu sulfate, Bordeaux mixture and 3 types of Cu oxychloride. Cu salts decreased soil pH, while this was not found with the other Cu sources. This difference in soil pH effects caused differences in the respiration, bacterial growth and fungal growth response. Basal soil respiration was negatively affected by Cu addition when the soil was spiked with Cu salts, but almost unaffected by commercial Cu fungicides. Bacterial growth was significantly and negatively affected by Cu addition for all the Cu sources, but Cu toxicity was higher for Cu salts than for commercial Cu fungicides. Fungal growth response was also different for Cu salts and commercial Cu fungicides, but only in the long-term. High Cu amendments using Cu salts stimulated fungal growth, whereas for commercial Cu fungicides, these concentrations inhibited fungal growth. Thus, the use of products similar to those used in commercial fungicides is a recommended practice for Cu risk assessments in soil.