Merck
  • Genomic and functional gene studies suggest a key role of beta-carotene oxygenase 1 like (bco1l) gene in salmon flesh color.
Genomic and functional gene studies suggest a key role of beta-carotene oxygenase 1 like (bco1l) gene in salmon flesh color.
Scientific reports (2019-12-29)
Hanna Helgeland, Marte Sodeland, Nina Zoric, Jacob Seilø Torgersen, Fabian Grammes, Johannes von Lintig, Thomas Moen, Sissel Kjøglum, Sigbjørn Lien, Dag Inge Våge
ABSTRACT
Red coloration of muscle tissue (flesh) is a unique trait in several salmonid genera, including Atlantic salmon. The color results from dietary carotenoids deposited in the flesh, whereas the color intensity is affected both by diet and genetic components. Herein we report on a genome-wide association study (GWAS) to identify genetic variation underlying this trait. Two SNPs on ssa26 showed strong associations to the flesh color in salmon. Two genes known to be involved in carotenoid metabolism were located in this QTL- region: beta-carotene oxygenase 1 (bco1) and beta-carotene oxygenase 1 like (bco1l). To determine whether flesh color variation is caused by one, or both, of these genes, functional studies were carried out including mRNA and protein expression in fish with red and pale flesh color. The catalytic abilities of these two genes were also tested with different carotenoids. Our results suggest bco1l to be the most likely gene to explain the flesh color variation observed in this population.