Merck
  • Preliminary Results, Perspectives, and Proposal for a Screening Method of In Vitro Susceptibility of Prototheca Species to Antimicrotubular Agents.
Preliminary Results, Perspectives, and Proposal for a Screening Method of In Vitro Susceptibility of Prototheca Species to Antimicrotubular Agents.
Antimicrobial agents and chemotherapy (2019-12-25)
Laura Morello, Tommaso Tiroli, Francesca Aretino, Stefano Morandi, Diego Breviario
ABSTRACT
Microorganisms belonging to the genus Prototheca are achlorophyllous microalgae, occasionally behaving as environmental pathogens that cause severe mastitis in milk cows, as well as localized or systemic infections in humans and animals. Among the different species belonging to the genus, Prototheca zopfii genotype 2 (recently reclassified as P. bovis) and P. blaschkeae are most commonly associated with bovine mastitis. To date, no pharmacological treatment is available to cure protothecal mastitis, and infected animals must be quarantined to avoid spreading the infection. The few antibiotic and antifungal drugs effective in vitro against Prototheca give poor results in vivo This failure is likely due to the lack of specificity of such drugs. As microalgae are more closely related to plants than to bacteria or fungi, an alternative possibility is to test molecules with herbicidal properties, in particular, antimicrotubular herbicides, for which plant rather than animal tubulin is the selective target. Once a suitable test protocol was set up, a panel of 11 antimicrotubular agents belonging to different chemical classes and selective for plant tubulin were tested for the ability to inhibit growth of Prototheca cells in vitro Two dinitroanilines, dinitramine and chloralin, showed strong inhibitory effects on P. blaschkeae at low micromolar concentrations, with half-maximal inhibitory concentrations (IC50) of 4.5 and 3 μM, respectively, while both P. zopfii genotype 1 (now reclassified as P. ciferrii) and P. bovis showed susceptibility to dinitramine only, to different degrees. Suitable screening protocols for antimitotic agents are suggested.