Merck
  • Fenfluramine diminishes NMDA receptor-mediated seizures via its mixed activity at serotonin 5HT2A and type 1 sigma receptors.
Fenfluramine diminishes NMDA receptor-mediated seizures via its mixed activity at serotonin 5HT2A and type 1 sigma receptors.
Oncotarget (2018-05-29)
María Rodríguez-Muñoz, Pilar Sánchez-Blázquez, Javier Garzón
ABSTRACT
Fenfluramine exhibits antiepileptic properties and thus diminishes epileptiform discharges in experimental animal models of Dravet syndrome. Fenfluramine is metabolized into norfenfluramine in vivo, which shows greater affinity and agonist activity at serotonin 5HT2 receptors (5HT2R) than fenfluramine. In this study, we found that fenfluramine and norfenfluramine disrupted the regulatory association of the sigma 1 receptor (σ1R) with NR1 subunits of glutamate N-methyl-D-aspartate receptors (NMDAR), an effect that was also produced by σ1R antagonists such as S1RA and prevented by σ1R agonists such as PPCC. The antagonists removed σ1R bound to NMDAR NR1 subunits enabling calcium-regulated calmodulin (CaM) to bind to those subunits. As a result, CaM may inhibit calcium permeation through NMDARs. The serotoninergic activity of fenfluramine at 5HT2AR, and likely also at 5HT2CR, collaborated with its activity at σ1Rs to prevent the convulsive syndrome promoted by NMDAR overactivation. Notably, fenfluramine enhanced the inhibitory coupling of G protein-coupled receptors such as 5HT1AR and cannabinoid type 1 receptor with NMDARs, thus allowing the more effective restrain of NMDAR activity. Thus, fenfluramine circumvents the negative side effects of direct NMDAR antagonists and may improve the quality of life of subjects affected by such proconvulsant dysfunctions.
Sigma-Aldrich
Anti-NMDAR2B Antibody, phosphoTyr 1472, Chemicon®, from rabbit
Sigma-Aldrich
Anti-NMDAR1 Antibody, (all splice variants), clone R1JHL, clone R1JHL, Chemicon®, from mouse