Merck
  • Simultaneous determination of uranium and thorium with Arsenazo III by second-derivative spectrophotometry.
Simultaneous determination of uranium and thorium with Arsenazo III by second-derivative spectrophotometry.
Talanta (1990-06-01)
R Kuroda, M Kurosaki, Y Hayashibe, S Ishimaru
ABSTRACT
A derivative spectrophotometric method has been developed for the simultaneous determination of microgram quantities of uranium and thorium with Arsenazo III in hydrochloric acid medium. The second-derivative absorbances of the uranium and thorium Arsenazo III complexes at 679.5 and 684.4 nm are used for their quantification. Uranium and thorium, both in the range 0.1-0.7 mug/ml have been determined simultaneously with good precision. The procedure does not require separation of uranium and thorium, and allows the determination of both metals in the presence of alkaline-earth metals and zirconium, but lanthanides interfere.