Merck
  • Design of Novel Haptens and Development of Monoclonal Antibody-Based Immunoassays for the Simultaneous Detection of Tylosin and Tilmicosin in Milk and Water Samples.
Design of Novel Haptens and Development of Monoclonal Antibody-Based Immunoassays for the Simultaneous Detection of Tylosin and Tilmicosin in Milk and Water Samples.
Biomolecules (2019-11-28)
Jian-Xin Huang, Chan-Yuan Yao, Jin-Yi Yang, Zhen-Feng Li, Fan He, Yuan-Xin Tian, Hong Wang, Zhen-Lin Xu, Yu-Dong Shen
ABSTRACT
In this work, a monoclonal antibody-based indirect competitive enzyme-linked immunosorbent assay (icELISA) was established to detect tylosin and tilmicosin in milk and water samples. A sensitive and specific monoclonal antibody was prepared by rational designed hapten, which was achieved by directly oxidizing the aldehyde group on the side chain of tylosin to the carboxyl group. Under the optimized conditions, the linear range of icELISA for tylosin and tilmicosin were 1.3 to 17.7 ng/mL and 2.0 to 47.4 ng/mL, with half-maximal inhibition concentration (IC50) values of 4.7 and 9.6 ng/mL, respectively. The cross-reactivity with other analogues of icELISA was less than 0.1%. The average recoveries of icELISA for tylosin and tilmicosin ranged from 76.4% to 109.5% in milk and water samples. Besides, the detection results of icELISA showed good correlations with HPLC-MS/MS. The proposed icELISA was satisfied for rapid and specific screening of tylosin and tilmicosin residues in milk and water samples.