Merck
  • APLNR promotes the progression of osteosarcoma by stimulating cell proliferation and invasion.
APLNR promotes the progression of osteosarcoma by stimulating cell proliferation and invasion.
Anti-cancer drugs (2019-03-29)
Lei Cui, Jing-Yuan Zhang, Zhi-Peng Ren, He-Jun Zhao, Gui-Shi Li
ABSTRACT
Osteosarcoma is the most common type of bone malignancies with a poor prognosis. In recent years, targeted therapy has shown great potential in the treatment of osteosarcoma, and more effective therapeutic targets for this disease need to be developed. APLNR is a seven transmembrane G-protein-coupled receptor expressed widely in multiple tissues. As has been reported, APLNR is involved in various physiological and pathological processes. Although APLNR plays a role in the development and progression of multiple tumors, the potential role of APLNR in osteosarcoma, a highly malignant tumor, remains unclear. Here, we reported that APLNR expression was correlated positively with clinical features including tumor size and stage of osteosarcoma. We found that APLNR knockdown inhibited the proliferation and invasion of osteosarcoma cells in vitro. In addition, APLNR could promote the progression and metastasis of osteosarcoma in mice. Collectively, this study showed the potential link between APLNR and osteosarcoma and suggested APLNR as a novel therapeutic target for osteosarcoma.