Merck
  • Cyclopropyl and isopropyl derivatives of 11-cis and 9-cis retinals at C-9 and C-13: subtle steric differences with major effects on ligand efficacy in rhodopsin.
Cyclopropyl and isopropyl derivatives of 11-cis and 9-cis retinals at C-9 and C-13: subtle steric differences with major effects on ligand efficacy in rhodopsin.
Journal of natural products (2011-02-12)
Willem J deGrip, Petra H M Bovee-Geurts, Yajie Wang, Michiel A Verhoeven, Johan Lugtenburg
ABSTRACT
Retinal is the natural ligand (chromophore) of the vertebrate rod visual pigment. It occurs in either the 11-cis (rhodopsin) or the 9-cis (isorhodopsin) configuration. In its evolution to a G protein coupled photoreceptor, rhodopsin has acquired exceptional photochemical properties. Illumination isomerizes the chromophore to the all-trans isomer, which acts as a full agonist. This process is extremely efficient, and there is abundant evidence that the C-9 and C-13 methyl groups of retinal play a pivotal role in this process. To examine the steric limits of the C-9 and C-13 methyl binding pocket of the binding site, we have prepared C-9 and C-13 cyclopropyl and isopropyl derivatives of its native ligands and of α-retinal at C-9. Most isopropyl analogues show very poor binding, except for 9-cis-13-isopropylretinal. Most cyclopropyl derivatives exhibit intermediate binding activity, except for 9-cis-13-cyclopropylretinal, which presents good binding activity. In general, the binding site shows preference for the 9-cis analogues over the 11-cis analogues. In fact, 13-isopropyl-9-cis-retinal acts as a superagonist after illumination. Another surprising finding was that 9-cyclopropylisorhodopsin is more like native rhodopsin with respect to spectral and photochemical properties, whereas 9-cyclopropylrhodopsin behaves more like native isorhodopsin in these aspects.