Merck
  • A BET Bromodomain Inhibitor Suppresses Adiposity-Associated Malignant Transformation.
A BET Bromodomain Inhibitor Suppresses Adiposity-Associated Malignant Transformation.
Cancer prevention research (Philadelphia, Pa.) (2017-12-17)
Debrup Chakraborty, Vanessa Benham, Vladislav Jdanov, Blair Bullard, Ana S Leal, Karen T Liby, Jamie J Bernard
ABSTRACT
Almost half a million of all new cancers have been attributed to obesity and epidemiologic evidence implicates visceral adipose tissue (VAT) and high-fat diets (HFD) in increasing cancer risk. We demonstrated that VAT-derived fibroblast growth factor 2 (FGF2) from mice fed an HFD or obese individuals stimulates the malignant transformation of epithelial cells. Mechanism-based strategies to prevent this VAT-enhanced tumorigenesis have not been explored. Clinical studies have indicated that bromodomain inhibitors have considerable potential as therapeutic agents for cancer by inhibiting the activity of several oncogenes, including c-Myc; however, their chemopreventive activity is unknown. We show herein that mice with visceral adiposity have elevated nuclear c-Myc expression in their epidermis. We hypothesized that the bromodomain inhibitor I-BET-762 (I-BET) would have efficacy in the prevention of malignant transformation by VAT and FGF2. We tested this hypothesis using our novel models of VAT-stimulated transformation in vitro and FGF2- stimulated tumor formation in vivo We found that I-BET significantly attenuates VAT and FGF2-stimulated transformation and inhibits VAT-induced c-Myc protein expression in several skin and breast epithelial cell lines. Moreover, I-BET attenuated tumor growth significantly in FGF2-treated nude mice. Work is ongoing to determine the role of visceral adiposity in c-Myc activity in several tissues and determine the inhibitory effect of I-BET on VAT-promoted tumors in vivoCancer Prev Res; 11(3); 129-42. ©2017 AACRSee related editorial by Berger and Scacheri, p. 125.