Merck
  • Preparation of soluble extracts from adenovirus-infected cells for studies of RNA splicing.
Preparation of soluble extracts from adenovirus-infected cells for studies of RNA splicing.
Methods in molecular medicine (2007-07-28)
Oliver Mühlemann, Göran Akusjärvi
ABSTRACT
Here we describe a collection of methods that have been adapted to produce highly efficient nuclear and cytoplasmic extracts from adenovirus-infected HeLa cells. We describe how to produce extracts from virus-infected cells and how to analyze RNA splicing in vitro using T7 RNA polymerase-derived splicing substrate RNAs.