Merck
  • miR-15a and miR-16 regulate serotonin transporter expression in human placental and rat brain raphe cells.
miR-15a and miR-16 regulate serotonin transporter expression in human placental and rat brain raphe cells.
The international journal of neuropsychopharmacology (2012-05-09)
Pablo R Moya, Jens R Wendland, Jennifer Salemme, Ruby L Fried, Dennis L Murphy
ABSTRACT
The serotonin transporter (SERT) is a key regulatory molecule in serotonergic transmission implicated in numerous biological processes relevant to human disorders. Recently, it was shown that SERT expression is controlled by miR-16 in mouse brain. Here, we show that SERT expression is regulated additionally by miR-15a as well as miR-16 in human and rat tissues. This post-transcriptional regulation was observed and characterized in reporter assays and likewise when endogenous SERT expression was evaluated in human placental choriocarcinoma JAR cells and rat brain raphe RN46A cells - two cell lines that endogenously express SERT. Similar effects for miR-16 to those of miR-15a were found in both human and rat cell lines. The effects of miR-15a and miR-16 were comparable in extent to those originally reported for miR-16 in mice. These findings represent a novel layer of complexity for SERT expression regulation exerted by the mir-15a/16 cluster, whose genes are adjacently located at human chromosome 13q14.3.