Merck
  • COA-Cl (2-Cl-C.OXT-A) can promote coronary collateral development following acute myocardial infarction in mice.
COA-Cl (2-Cl-C.OXT-A) can promote coronary collateral development following acute myocardial infarction in mice.
Scientific reports (2019-02-24)
Toshiyuki Nishikido, Jun-Ichi Oyama, Aya Shiraki, Ikuko Tsukamoto, Junsuke Igarashi, Koichi Node
ABSTRACT
2-Cl-C.OXT-A (COA-Cl) is a novel nucleic acid analogue that promotes tube-forming activity of human umbilical vein endothelial cells (HUVEC) through vascular endothelial growth factor (VEGF). The development of coronary collateral circulation is critical to rescue the ischemic myocardium and to prevent subsequent irreversible ischemic injury. We evaluated whether COA-Cl can promote angiogenesis in ischemic tissue, reduce infarct size and preserve cardiac contractility in vivo. Mice received COA-Cl or placebo daily for three days after myocardial infarction (MI) by coronary ligation. The degree of angiogenesis in ischemic myocardium was assessed by staining endothelial cells and vascular smooth muscle cells, and measuring infarct size/area-at-risk. In mice treated with COA-Cl, enhanced angiogenesis and smaller infarct size were recognized, even given a similar area at risk. We observed increases in the protein expression levels of VEGF and in the protein phosphorylation level of eNOS. In addition, the heart weight to body weight ratio and myocardial fibrosis in COA-Cl mice were decreased on Day 7. Administration of COA-Cl after MI promotes angiogenesis, which is associated with reduced infarct size and attenuated cardiac remodeling. This may help to prevent heart failure due to cardiac dysfunction after MI.