Merck
  • Trained Memory of Human Uterine NK Cells Enhances Their Function in Subsequent Pregnancies.
Trained Memory of Human Uterine NK Cells Enhances Their Function in Subsequent Pregnancies.
Immunity (2018-05-17)
Moriya Gamliel, Debra Goldman-Wohl, Batya Isaacson, Chamutal Gur, Natan Stein, Rachel Yamin, Michael Berger, Myriam Grunewald, Eli Keshet, Yoach Rais, Chamutal Bornstein, Eyal David, Adam Jelinski, Iris Eisenberg, Caryn Greenfield, Arbel Ben-David, Tal Imbar, Ronit Gilad, Ronit Haimov-Kochman, David Mankuta, Matan Elami-Suzin, Ido Amit, Jacob H Hanna, Simcha Yagel, Ofer Mandelboim
ABSTRACT
Natural killer cells (NKs) are abundant in the human decidua, regulating trophoblast invasion and angiogenesis. Several diseases of poor placental development are associated with first pregnancies, so we thus looked to characterize differences in decidual NKs (dNKs) in first versus repeated pregnancies. We discovered a population found in repeated pregnancies, which has a unique transcriptome and epigenetic signature, and is characterized by high expression of the receptors NKG2C and LILRB1. We named these cells Pregnancy Trained decidual NK cells (PTdNKs). PTdNKs have open chromatin around the enhancers of IFNG and VEGFA. Activation of PTdNKs led to increased production and secretion of IFN-γ and VEGFα, with the latter supporting vascular sprouting and tumor growth. The precursors of PTdNKs seem to be found in the endometrium. Because repeated pregnancies are associated with improved placentation, we propose that PTdNKs, which are present primarily in repeated pregnancies, might be involved in proper placentation.