Merck
  • Testis cord differentiation after the sex determination stage is independent of Sox9 but fails in the combined absence of Sox9 and Sox8.
Testis cord differentiation after the sex determination stage is independent of Sox9 but fails in the combined absence of Sox9 and Sox8.
Developmental biology (2009-01-07)
Francisco Barrionuevo, Ina Georg, Harry Scherthan, Charlotte Lécureuil, Florian Guillou, Michael Wegner, Gerd Scherer
ABSTRACT
Sox9 and Sox8 are transcription factors expressed in embryonic and postnatal Sertoli cells of the mouse testis. Sox9 inactivation prior to the sex determination stage leads to complete XY sex reversal. In contrast, there is normal embryonic testis development in Sox8 mutants which are initially fertile, but later develop progressive seminiferous tubule failure and infertility. To determine whether Sox9 is required for testis development after the initial steps of sex determination, we crossed Sox9(flox) mice with an AMH-Cre transgenic line thereby completely deleting Sox9 in Sertoli cells by E14.0. Conditional Sox9 null mutants show normal embryonic testis development and are initially fertile, but, like Sox8(-/-) mutants, become sterile from dysfunctional spermatogenesis at about 5 months. To see whether Sox8 may compensate for the absence of Sox9 during embryonic testis differentiation, we generated a Sox9 conditional knockout on a Sox8 mutant background. In the double mutants, differentiation of testis cords into seminiferous testis tubules ceases after P6 in the absence of one Sox8 allele, and after P0 in the absence of both Sox8 alleles, leading to complete primary infertility. Sox9,Sox8 double nullizygous testes show upregulation of early ovary-specific markers and downregulation of Sertoli intercellular junctions at E15.5. Their very low Amh levels still cause complete regression of the Müllerian duct but with reduced penetrance. This study shows that testis cord differentiation is independent of Sox9, and that concerted Sox9 and Sox8 function in post E14.0 Sertoli cells is essential for the maintenance of testicular function.
Sigma-Aldrich
Anti-Neural Cell Adhesion Molecule Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse