Merck
  • FLI-06 Intercepts Notch Signaling And Suppresses The Proliferation And Self-renewal Of Tongue Cancer Cells.
FLI-06 Intercepts Notch Signaling And Suppresses The Proliferation And Self-renewal Of Tongue Cancer Cells.
OncoTargets and therapy (2019-10-02)
Rui-Huan Gan, Li-Song Lin, Jing Xie, Li Huang, Lin-Can Ding, Bo-Hua Su, Xian-E Peng, Da-Li Zheng, You-Guang Lu
ABSTRACT
The Notch signaling pathway plays an oncogenic role in tongue squamous cell carcinoma. The aim of this study was to inhibit the proliferation and self-renewal of tongue cancer cells by applying Notch signaling pathway inhibitor FLI-06 (Selleck, USA) and to lay a foundation for the clinically targeted treatment of tongue cancer for the future. The mRNA expression level of Notch1 and the overall survival rate of patients with tongue cancer were examined by analyzing the TCGA database. Tongue cancer cells were treated with FLI-06. Cell proliferation, apoptosis, and stem cell self-renewal ability were tested in appropriate ways. A xenograft mouse model was established to observe tumor growth. From the TCGA data, we demonstrated that patients with high expression of Notch1 had a poor prognosis. We observed that the Notch signaling pathway inhibitor FLI-06 can restrain the activation of the Notch signaling pathway, decrease cell proliferation and induce cell apoptosis in vitro. The xenograft experiment indicated that intraperitoneal injection of FLI-06 inhibited tumor growth and increased cell apoptosis. FLI-06 suppressed both the mRNA and protein expression of Notch receptor and Notch targeted genes. We also observed that FLI-06 suppressed the proliferation of tongue cancer stem cells. FLI-06 can block the proliferation and self-renewal of tongue cancer cells. It is inferred that this compound, which inhibits the Notch signaling pathway, may serve as a potential targeted drug for the treatment of tongue cancer in the clinic.