Merck
  • Nucleoside supplementation modulates mitochondrial DNA copy number in the dguok -/- zebrafish.
Nucleoside supplementation modulates mitochondrial DNA copy number in the dguok -/- zebrafish.
Human molecular genetics (2018-11-15)
Benjamin Munro, Rita Horvath, Juliane S Müller
ABSTRACT
Deoxyguanosine kinase (dGK) is an essential rate-limiting component of the mitochondrial purine nucleotide salvage pathway, encoded by the nuclear gene encoding deoxyguanosine kinase (DGUOK). Mutations in DGUOK lead to mitochondrial DNA (mtDNA) depletion typically in the liver and brain, causing a hepatocerebral phenotype. Previous work has shown that in cultured DGUOK patient cells it is possible to rescue mtDNA depletion by increasing substrate amounts for dGK. In this study we developed a mutant dguok zebrafish (Danio rerio) line using CRISPR/Cas9 mediated mutagenesis; dguok-/- fish have significantly reduced mtDNA levels compared with wild-type (wt) fish. When supplemented with only one purine nucleoside (dGuo), mtDNA copy number in both mutant and wt juvenile animals was significantly reduced, contrasting with previous cell culture studies, possibly because of nucleotide pool imbalance. However, in adult dguok-/- fish we detected a significant increase in liver mtDNA copy number when supplemented with both purine nucleosides. This study further supports the idea that nucleoside supplementation has a potential therapeutic benefit in mtDNA depletion syndromes by substrate enhancement of the purine nucleoside salvage pathway and might improve the liver pathology in patients.