Merck
The anticonvulsant MK-801 is a potent N-methyl-D-aspartate antagonist.
Proceedings of the National Academy of Sciences of the United States of America (1986-09-01)
E H Wong, J A Kemp, T Priestley, A R Knight, G N Woodruff, L L Iversen
ABSTRACT
The compound MK-801 [(+)-5-methyl-10,11-dihydro-5H-dibenzo[a,d] cyclohepten-5,10-imine maleate)] is a potent anticonvulsant that is active after oral administration and whose mechanism of action is unknown. We have detected high-affinity (Kd = 37.2 +/- 2.7 nM) binding sites for [3H]MK-801 in rat brain membranes. These sites are heat-labile, stereoselective, and regionally specific, with the hippocampus showing the highest density of sites, followed by cerebral cortex, corpus striatum, and medulla-pons. There was no detectable binding in the cerebellum. MK-801 binding sites exhibited a novel pharmacological profile, since none of the major neurotransmitter candidates were active at these sites. The only compounds that were able to compete for [3H]MK-801 binding sites were substances known to block the responses of excitatory amino acids mediated by the N-methyl-D-aspartate (N-Me-D-Asp) receptor subtype. These comprised the dissociative anesthetics phencyclidine and ketamine and the sigma-type opioid N-allylnormetazocine (SKF 10,047). Neurophysiological studies in vitro, using a rat cortical-slice preparation, demonstrated a potent, selective, and noncompetitive antagonistic action of MK-801 on depolarizing responses to N-Me-D-Asp but not to kainate or quisqualate. The potencies of phencyclidine, ketamine, SKF 10,047, and the enantiomers of MK-801 as N-Me-D-Asp antagonists correlated closely (r = 0.99) with their potencies as inhibitors of [3H]MK-801 binding. This suggests that the MK-801 binding sites are associated with N-Me-D-Asp receptors and provides an explanation for the mechanism of action of MK-801 as an anticonvulsant.