Merck
  • Intratumoral delivery of low doses of anti-CD40 mAb combined with monophosphoryl lipid a induces local and systemic antitumor effects in immunocompetent and T cell-deficient mice.
Intratumoral delivery of low doses of anti-CD40 mAb combined with monophosphoryl lipid a induces local and systemic antitumor effects in immunocompetent and T cell-deficient mice.
Journal of immunotherapy (Hagerstown, Md. : 1997) (2012-12-06)
Tyler J Van De Voort, Mildred A R Felder, Richard K Yang, Paul M Sondel, Alexander L Rakhmilevich
ABSTRACT
In this study, an agonistic anti-CD40 monoclonal antibody was combined with monophosphoryl lipid A (MPL), a nontoxic derivative of lipopolysaccharide and agonist of toll-like receptor-4, to assess the immunomodulatory and antitumor synergy between the 2 agents in mice. Anti-CD40 was capable of priming macrophages to subsequent ex vivo activation by MPL in immunocompetent and T-cell-depleted mice. Intraperitoneal injections of anti-CD40+MPL induced additive to synergistic suppression of poorly immunogenic B16-F10 melanoma growing subcutaneously in syngeneic mice. When anti-CD40+MPL were injected directly into the subcutaneous tumor, the combination treatment was more effective, even with a 25-fold reduction in dose. Low-dose intratumoral treatment also slowed the growth of a secondary tumor growing simultaneously at a distant, untreated site. Antitumor effects were also induced in severe combined immunodeficiency mice and in T-cell-depleted C57BL/6 mice. Taken together, our results show that the antitumor effects of anti-CD40 are enhanced by subsequent treatment with MPL, even in T-cell-deficient hosts. These preclinical data suggest that an anti-CD40+MPL combined regimen is appropriate for clinical testing in human patients, including cancer patients who may be immunosuppressed from prior chemotherapy.