Merck
  • 3-Deoxysappanchalcone inhibits tumor necrosis factor-α-induced matrix metalloproteinase-9 expression in human keratinocytes through activated protein-1 inhibition and nuclear factor-kappa B DNA binding activity.
3-Deoxysappanchalcone inhibits tumor necrosis factor-α-induced matrix metalloproteinase-9 expression in human keratinocytes through activated protein-1 inhibition and nuclear factor-kappa B DNA binding activity.
Biological & pharmaceutical bulletin (2011-06-02)
Ui Joung Youn, Kung-Woo Nam, Hui-Seong Kim, Goya Choi, Woo Seok Jeong, Mi Young Lee, Sungwook Chae
ABSTRACT
Tumor necrosis factor α (TNF-α), which is a primary cytokine responsible for inflammatory responses in skin, induces the synthesis of matrix metalloproteinase-9 (MMP-9), which causes skin aging. The protective effects of 3-deoxysappanchalcone against TNF-α-induced damage was investigated using human skin keratinocytes. The results showed that 3-deoxysappanchalcone inhibited MMP-9 expression at the protein and mRNA level, by blocking the activation of activator protein-1 (AP-1) and nuclear factor kappa B (NF-κB). Taken together, the inhibitory activity of 3-deoxysappanchalcone on MMP-9 expression and production in TNF-α-treated cells was found to be mediated by the suppression of AP-1 and NF-κB activation.