Merck
  • Estimating the risk of drug-induced proarrhythmia using human induced pluripotent stem cell-derived cardiomyocytes.
Estimating the risk of drug-induced proarrhythmia using human induced pluripotent stem cell-derived cardiomyocytes.
Toxicological sciences : an official journal of the Society of Toxicology (2011-06-23)
Liang Guo, Rory M C Abrams, Joshua E Babiarz, Jennifer D Cohen, Sei Kameoka, Martin J Sanders, Eric Chiao, Kyle L Kolaja
ABSTRACT
Improved in vitro systems for predicting drug-induced toxicity are needed in the pharmaceutical and biotechnology industries to decrease late-stage drug attrition. One unmet need is an early screen for cardiotoxicity, which accounts for about one third of safety-based withdrawn pharmaceuticals. Herein, the first published report of a high-throughput functional assay employing a monolayer of beating human induced pluripotent stem cell-derived cardiomyocytes (iPSC-CMs) is described, detailing a model that accurately detects drug-induced cardiac abnormalities. Using 96-well plates with interdigitated electrode arrays that assess impedance, the rhythmic, synchronous contractions of the iPSC-CMs were detected. Treatment of the iPSC-CMs with 28 different compounds with known cardiac effects resulted in compound-specific changes in the beat rate and/or the amplitude of the impedance measurement. Changes in impedance for the compounds tested were comparable with the results from a related technology, electric field potential assessment obtained from microelectrode arrays. Using the results from the set of compounds, an index of drug-induced arrhythmias was calculated, enabling the determination of a drug's proarrhythmic potential. This system of interrogating human cardiac function in vitro opens new opportunities for predicting cardiac toxicity and studying cardiac biology.