Merck
  • miR-5590-3p-YY1 feedback loop promotes the proliferation and migration of triple-negative breast cancer cells.
miR-5590-3p-YY1 feedback loop promotes the proliferation and migration of triple-negative breast cancer cells.
Journal of cellular biochemistry (2019-06-14)
Feng Liang, Xin Fu, Linlin Wang
ABSTRACT
Lacking of diagnostic and prognostic biomarkers is a significant reason for the poor prognosis of patients with triple-negative breast cancer (TNBC). MicroRNAs (miRNAs) have been discovered to engage in the tumorigenesis and development of TNBC. miR-5590-3p has been found to be involved in the development of gastric cancer, but its role and underlying mechanism in TNBC remain obscure. In this study, it was discovered that miR-5590-3p was downregulated in TNBC tissues and cells. Function assays confirmed that miR-5590-3p overexpression inhibited cell proliferation, migration, and epithelial-mesenchymal transition (EMT) process as well as promoted cell apoptosis in TNBC. Moreover, YY1 could bind with the promoter of miR-5590-3p and overexpression of YY1 inhibited the transcription of miR-5590-3p. It was found that YY1 acted as a downstream target gene to bind with miR-5590-3p and was negatively regulated by miR-5590-3p. Finally, it was discovered that overexpression of YY1 could partially rescue the miR-5590-3p overexpression-mediated inhibitive effect on TNBC progression. Taken together these results, it can be concluded that miR-5590-3p-YY1 feedback loop promoted the proliferation and migration of TNBC.