Merck
  • Glucocorticoids regulate pentraxin-3 expression in human airway smooth muscle cells.
Glucocorticoids regulate pentraxin-3 expression in human airway smooth muscle cells.
PloS one (2019-08-23)
Jingbo Zhang, Latifa Koussih, Lianyu Shan, Andrew J Halayko, Omar Tliba, Abdelilah S Gounni
ABSTRACT
Pentraxin-3 (PTX3) is a multifunctional protein involved in both innate and adaptive immunity. Glucocorticoid (GC) is the first-line therapy to mitigate airway inflammation in asthma. Previous pieces of evidence showed that GC has divergent effects on PTX3 production in various cell types. The molecular mechanisms controlling PTX3 expression in HASMC are, however, not yet characterized. In this study, we demonstrate that the synthetic GC, dexamethasone (DEX) increases the expression of PTX3 both at the protein and mRNA levels. We also found that such an effect of DEX was dependent on de novo protein synthesis and the GC receptor (GR). While DEX increases PTX3 mRNA stability, it did not affect its promoter activity. Interestingly, HASMC pre-treated with p42/p44 ERK inhibitor, but not with p38 or JNK-MAPK inhibitors, significantly interfered with DEX-induced PTX3 secretion. Taken together, our data suggest that GC regulates PTX3 expression in HASMC through transcriptional and post-transcriptional mechanisms in a GR and ERK-dependent manner.
Sigma-Aldrich
Mouse Ptx3 / Pentraxin-related Protein PTX3 ELISA Kit