Merck
  • Phenotypic and functional changes in gamma delta T lymphocytes from HTLV-1 carriers.
Phenotypic and functional changes in gamma delta T lymphocytes from HTLV-1 carriers.
Journal of leukocyte biology (2019-07-10)
Raquel Cavalcanti De Albuquerque, Alessandra Granato, Isabela Silva Castro, Rafael Carvalho Torres, Flávia Santos Souza, Marco Antonio Lima, Ana Claudia Celestino Bezerra Leite, Otávio de Melo Espíndola, Juliana Echevarria-Lima
ABSTRACT
Human T-cell lymphotropic virus type-1 (HTLV-1) is the etiologic agent of HTLV-1-associated myelopathy/tropical spastic paraparesis (HAM/TSP), which is a chronic inflammatory disease that leads to gradual loss of motor movement as a result of the death of spinal cord cells through immune mediated mechanisms. The risk to develop HAM/TSP disease positively correlates with the magnitude of HTLV-1 proviral load. Gamma-delta T lymphocytes have been recognized as important players in a variety of infectious diseases. Therefore, we have investigated interactions between HTLV-1 infection and γδ T lymphocytes during HAM/TSP. Similar frequencies of total γδ T lymphocytes and their Vγ9δ2+ and Vγ9δ2neg subpopulations were observed in HAM/TSP patients. However, T lymphocytes obtained from HTLV-1 carriers displayed significantly higher rates of spontaneous proliferation and NKp30 expression when compared to cells from uninfected donors. In addition, an important decrease in the frequency of granzyme B+ γδ T lymphocytes (approximately 50%) was observed in HAM/TSP patients. Higher proportion of IFN-γ+ γδ T lymphocytes was found in HTLV-1-infected patients, which positively correlated with the HTLV-1 proviral load in peripheral blood mononuclear cells. Collectively, our data indicates that HTLV-1 infection leads to phenotypic and functional changes in the population of γδ T lymphocyte population, suggesting that HTLV-1 infection modulates functions associated to these cells, which might be involved in controlling the infection or in the development of HTLV-1-associated diseases.