Merck
  • Language-Related White-Matter-Tract Deficits in Children with Benign Epilepsy with Centrotemporal Spikes: A Retrospective Study.
Language-Related White-Matter-Tract Deficits in Children with Benign Epilepsy with Centrotemporal Spikes: A Retrospective Study.
Journal of clinical neurology (Seoul, Korea) (2019-10-09)
Hyun Ho Kim, Gyung Ho Chung, Sung Hee Park, Sun Jun Kim
ABSTRACT
Benign epilepsy with centrotemporal spikes (BECTS) is one of the most common pediatric epilepsies, and it generally has a good prognosis. However, recent research has indicated that the epileptic activity of BECTS can cause cognitive defects such as language, visuospatial, and auditory verbal memory deficits. This study assessed language-delivery deficits in BECTS patients using diffusion-tensor magnetic resonance imaging (DTI). T1-weighted MRI, DTI, and language tests were conducted in 16 BECTS patients and 16 age-matched controls. DTI data were analyzed using the TRActs Constrained by Underlying Anatomy tool in FreeSurfer 5.3, and 18 major white-matter tracts were extracted, which included 4 language-related tracts: the inferior longitudinal fasciculus, superior longitudinal fasciculus-parietal terminations, superior longitudinal fasciculus-temporal terminations, and uncinate fasciculus (UNC). Language tests included the Korean version of the Receptive and Expressive Vocabulary Test, Test of Problem-Solving Abilities (TOPS), and the mean length of utterance in words. The BECTS group exhibited decreased mean fractional anisotropy and increased mean radial diffusivity, with significant differences in both the superior longitudinal fasciculus and the left UNC (p<0.05), which are the language-related white-matter tracts in the dual-loop model. The TOPS language test scores were significantly lower in the BECTS group than in the control group (p<0.05). It appears that BECTS patients can exhibit language deficits. Seizure activities of BECTS could alter DTI scalar values in the language-related white-matter tracts.