Merck
  • Implication of Kv7 Channels in the Spinal Antinociceptive Actions of Celecoxib.
Implication of Kv7 Channels in the Spinal Antinociceptive Actions of Celecoxib.
The Journal of pharmacology and experimental therapeutics (2019-06-27)
Jorge Vicente-Baz, Jose A Lopez-Garcia, Ivan Rivera-Arconada
ABSTRACT
Celecoxib is a nonsteroidal anti-inflammatory drug (NSAID) commonly used to treat pain conditions in humans. In addition to its blocking activity on cyclooxygenase (COX) enzymes, several other targets could contribute to its analgesic activity. Here we explore the spinal antinociceptive actions of celecoxib and the potential implication of Kv7 channels in mediating its effects. Spinal cord in vitro preparations from hind paw-inflamed animals were used to assess the segmental sensory-motor and the early sensory processing of nociceptive information. Electrophysiological recordings of ventral roots and dorsal horn neurones were obtained, and the effects of celecoxib and Kv7 modulators on responses to repetitive dorsal root stimulation at C-fiber intensity were assessed. Celecoxib applied at clinically relevant concentrations produced depressant effects on responses to dorsal root stimulation recorded from both ventral roots and individual dorsal horn neurones; by contrast, the non-nociceptive monosynaptic reflex was unaffected. The NSAID indomethacin had no effect on spinal reflexes, but further coapplication of celecoxib still produced depressant effects. The depressant actions of celecoxib were abolished after Kv7 channel blockade and mimicked by its structural analog dimethyl-celecoxib, which lacks COX-blocking activity. The present results identify Kv7 channels as novel central targets for celecoxib, which may be relevant to its analgesic effect. This finding contributes to better understand the pharmacology of celecoxib and reinforces both the role of Kv7 channels in modulating the excitability of central pain pathways and its validity as target for the design of analgesics.