Merck
  • Inhibition of angiogenesis and breast cancer in mice by the microtubule inhibitors 2-methoxyestradiol and taxol.
Inhibition of angiogenesis and breast cancer in mice by the microtubule inhibitors 2-methoxyestradiol and taxol.
Cancer research (1997-01-01)
N Klauber, S Parangi, E Flynn, E Hamel, R J D'Amato
ABSTRACT
2-Methoxyestradiol (2-ME), an endogenous estrogen metabolite which disrupts microtubule function, has been shown to inhibit proliferating cells in vitro and suppress certain murine tumors in vivo. In vitro screening has determined that breast cancer cell lines are most sensitive to inhibition by 2-ME. Additionally, 2-ME has been shown to inhibit angiogenesis in vitro. We tested whether 2-ME suppresses cytokine-induced angiogenesis in vivo and inhibits growth of a human breast carcinoma in severe combined immunodeficient mice. A model of basic fibroblast growth factor (bFGF) and vascular endothelial growth factor (VEGF)-induced corneal neovascularization in C57BL/6 mice was used to evaluate the antiangiogenic effects of 2-ME and other microtubule inhibitors such as Taxol, vincristine, and colchicine. 2-ME (150 mg/kg p.o., n = 20) inhibited bFGF and VEGF-induced neovascularization by 39% and 54%, respectively. Taxol (6 mg/kg i.p., n = 17) inhibited bFGF and VEGF-induced neovascularization by 45% and 37%, respectively. Vincristine (0.2 mg/kg i.p., n = 8) and colchicine (0.25 mg/kg i.p., n = 8) had no effect. Treatment with 2-ME (75 mg/kg p.o., n = 9) for 1 month suppressed the growth of a human breast carcinoma in mice by 60% without toxicity. Recognition of the antiangiogenic and antitumor properties of 2-ME and Taxol may be crucial in planning clinical applications to angiogenesis-dependent diseases.