Merck
  • Caspase-4 mediates cytoplasmic accumulation of TDP-43 in the primate brains.
Caspase-4 mediates cytoplasmic accumulation of TDP-43 in the primate brains.
Acta neuropathologica (2019-02-28)
Peng Yin, Xiangyu Guo, Weili Yang, Sen Yan, Su Yang, Ting Zhao, Qiang Sun, Yunbo Liu, Shihua Li, Xiao-Jiang Li
ABSTRACT
The cytoplasmic accumulation of the nuclear TAR DNA-binding protein 43 (TDP-43) is a pathologic hallmark in amyotrophic lateral sclerosis, frontotemporal lobar degeneration, and other neurological disorders. However, most transgenic TDP-43 rodent models show predominant nuclear distribution of TDP-43 in the brain. By expressing mutant TDP-43 (M337V) in the brains of rhesus monkeys and mice, we verified that mutant TDP-43 is distributed in the cytoplasm of the monkey brain and that the majority of mutant TDP-43 remains in the nuclei of the mouse brain. The primate-specific caspase-4, but not mouse homologue caspase-11, could remove the NLS-containing N-terminal domain and generate fragmented TDP-43 that accumulates in the cytoplasm. Moreover, increased expression of caspase-4 in the monkey brain promotes the cytoplasmic accumulation of endogenous TDP-43, and suppressing caspase-4 reduces the cytoplasmic distribution of endogenous TDP-43 in cultured human neural cells. Our findings suggest that primate-specific caspase-4-mediated cleavage of TDP-43 accounts for its cytoplasmic mislocalization in the primate brains and may serve as a potential therapeutic target.
Sigma-Aldrich
Anti-phospho TDP-43 (Ser409/Ser410) Antibody, clone 1D3, clone 1D3, from rat
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein (GFAP) Antibody, serum, Chemicon®