Merck
Galectin-1 is a new fibrosis protein in type 1 and type 2 diabetes.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2018-07-06)
Noor Al-Obaidi, Sumathy Mohan, Sitai Liang, Zhenze Zhao, Bijaya K Nayak, Boajie Li, P Sriramarao, Samy L Habib
ABSTRACT
Chronic exposure of tubular renal cells to high glucose contributes to tubulointerstitial changes in diabetic nephropathy. In the present study, we identified a new fibrosis gene called galectin-1 (Gal-1), which is highly expressed in tubular cells of kidneys of type 1 and type 2 diabetic mouse models. Gal-1 protein and mRNA expression showed significant increase in kidney cortex of heterozygous Akita+/- and db/db mice compared with wild-type mice. Mouse proximal tubular cells exposed to high glucose showed significant increase in phosphorylation of Akt and Gal-1. We cloned Gal-1 promoter and identified the transcription factor AP4 as binding to the Gal-1 promoter to up-regulate its function. Transfection of cells with plasmid carrying mutations in the binding sites of AP4 to Gal-1 promoter resulted in decreased protein function of Gal-1. In addition, inhibition of Gal-1 by OTX-008 showed significant decrease in p-Akt/AP4 and protein-promoter activity of Gal-1 and fibronectin. Moreover, down-regulation of AP4 by small interfering RNA resulted in a significant decrease in protein expression and promoter activity of Gal-1. We found that kidney of Gal-1-/- mice express very low levels of fibronectin protein. In summary, Gal-1 is highly expressed in kidneys of type 1 and 2 diabetic mice, and AP4 is a major transcription factor that activates Gal-1 under hyperglycemia. Inhibition of Gal-1 by OTX-008 blocks activation of Akt and prevents accumulation of Gal-1, suggesting a novel role of Gal-1 inhibitor as a possible therapeutic target to treat renal fibrosis in diabetes.-Al-Obaidi, N., Mohan, S., Liang, S., Zhao, Z., Nayak, B. K., Li, B., Sriramarao, P., Habib, S. L. Galectin-1 is a new fibrosis protein in type 1 and type 2 diabetes.