Merck
  • Inhibition of rat liver arginase by an intermediate in NO biosynthesis, NG-hydroxy-L-arginine: implications for the regulation of nitric oxide biosynthesis by arginase.
Inhibition of rat liver arginase by an intermediate in NO biosynthesis, NG-hydroxy-L-arginine: implications for the regulation of nitric oxide biosynthesis by arginase.
Biochemical and biophysical research communications (1994-07-15)
F Daghigh, J M Fukuto, D E Ash
ABSTRACT
NG-hydroxy-L-arginine, an intermediate in the biosynthesis of nitric oxide (NO), has been found to be a uniquely potent competitive inhibitor of rat liver arginase. Among previously reported inhibitors of arginase and the eight arginine analogs tested herein, only NG-hydroxy-L-arginine was found to be strongly inhibitory. Significantly, the Ki (42 microM) for inhibition of rat liver arginase by NG-hydroxy-L-arginine was found to be 20-40-fold lower than the KM (1-1.7 mM) for its natural substrate, L-arginine. Since NG-hydroxy-L-arginine is the only known intermediate in the biosynthesis of NO from L-arginine, this finding may have significant implications for the regulation of NO levels in tissues or cells, such as liver or macrophages, which synthesize both NO and contain arginase.