- Purification and characterization of two extracellular alkaline phosphatases from a psychrophilic arthrobacter isolate.
Purification and characterization of two extracellular alkaline phosphatases from a psychrophilic arthrobacter isolate.
Applied and environmental microbiology (1997-07-01)
P De Prada, J E Brenchley
PMID16535659
ABSTRACT
Two extracellular, heat-labile alkaline phosphatases were purified from a psychrophilic Arthrobacter isolate, D10. The enzymes were active over different pH ranges, used distinct substrates, and had different kinetic properties. Each enzyme reacted specifically to its own antibody during immunoblot analysis. One had both monophosphatase and diesterase activities.