Merck
  • Purification and partial characterization of a novel phosphodiesterase from the venom of Trimeresurus stejnegeri: inhibition of platelet aggregation.
Purification and partial characterization of a novel phosphodiesterase from the venom of Trimeresurus stejnegeri: inhibition of platelet aggregation.
Biochimie (2011-06-15)
Lili Peng, Xiaolong Xu, Dengke Shen, Yan Zhang, Jiajia Song, Xincheng Yan, Mingchun Guo
ABSTRACT
The phosphodiesterases (PDEs) are a superfamily of enzymes that have multiple roles in extracellular nucleotide metabolism and in the regulation of nucleotide-based intercellular signaling. Here we describe for the first time the isolation and partial characterization of a novel phosphodiesterase from Trimeresurus stejnegeri venom, named TS-PDE, using ion exchange and gel filtration chromatography. The purified TS-PDE is shown to be homogeneous as judged by SDS-PAGE and capillary isoelectric focusing. TS-PDE is a glycoprotein which contains 2.48% carbohydrate. Unlike other PDEs which are usually single polypeptide chain proteins with isoelectric points between 7.5 and 10.5, TS-PDE is a disulfide-linked heterodimer with an isoelectric point of 5.1 and a molecular mass of 100 kDa. The N-terminal amino acids of two chains are valine and serine, respectively. Furthermore, among all identified PDEs, only TS-PDE contains both of endogenous Cu(2+) and Zn(2+) which are essential for its phosphodiesterase activity. The purified TS-PDE exhibits broad phosphodiesterase substrate range with the order of specificity: nicotinamide guanine dinucleotide > ATP > nicotinamide adenine dinucleotide > ADP. The purified TS-PDE shows an exonuclease activity and no contamination with either alkaline phosphatase or 5'-nucleotidase activity. TS-PDE strongly inhibits ADP-induced platelet aggregation in human platelet-rich plasma by hydrolyzing ADP. Altogether, these results indicate that the novel TS-PDE is a unique phosphodiesterase with different structure from the known PDEs.