Merck
  • High level expression, purification and activation of human dipeptidyl peptidase I from mammalian cells.
High level expression, purification and activation of human dipeptidyl peptidase I from mammalian cells.
Protein expression and purification (2010-09-11)
Wei Yang, Wenjuan Xia, Jingjing Mao, Daqi Xu, Jianhe Chen, Shan Feng, Jianhua Wang, Hua Li, Claus Friis Theisen, Jørn Meidahl Petersen, Matthías Thórólfsson, Hanne Benedicte Rasmussen, Flemming Junker, Esper Boel, Jing Su
ABSTRACT
Dipeptidyl peptidase I (DPPI) plays a crucial role in maturation of many regulatory peptides and has been suggested as a pharmaceutical target in several inflammatory diseases. It is also a useful processing enzyme for the generation of authentic protein products by catalyzing the removal of N-terminal fusion peptides. We used a robust transient transfection system in human embryonic kidney 293 cells to exploit expression and activation of DPPI from chicken, rat and man for the development of an industrial production process. The expression of human and rat DPPI was significantly higher in the human HEK293 cell line than that obtained with avian DPPI. A CHO K1SV stable cell line was selected as the optimal stable host system for production of human DPPI yielding expression levels higher than 1.5 g/L. The secreted pro-DPPI underwent auto-maturation during defined buffer conditions during the purification steps. Active human DPPI was purified with a three-step purification strategy employing: Butyl Sepharose 4 Fast Flow, Sephadex G-25 Medium and Q Sepharose Fast Flow chromatography. The final yield of active enzyme was approximately 1 g/L cell culture. The enzyme exhibited exopeptidase activity against both a dipeptide-p-nitroanilide substrate and N-terminally extended MEAE-hGH (Met-Glu-Ala-Glu-human growth hormone). In conclusion, an efficient production process for recombinant human DPPI has been developed including a highly efficient and stable CHO cell system and an efficient purification procedure, which is simple and easy to scale for industrial purposes. The present data facilitates not only industrial applications of DPPI as a processing enzyme, but also provides active enzyme useful in the identification of small molecule inhibitors.